Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Medium, Blue, 14-6N34EC, 100/Box
Catalog No. 503043613
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit The medium size McKesson powder-free nitrile exam gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up and becoming loose The powder-free medium nitrile exam gloves are free of the allergen natural rubber latex
Catalog No. 50-304-3613 Supplier Med Vet International Supplier No. 1107941BX
Exam Glove McKesson Confiderm 3.0 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit The medium size McKesson powder-free nitrile exam gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up and becoming loose The powder-free medium nitrile exam gloves are free of the allergen natural rubber latex