Learn More
Med Vet International McKesson Pink Nitrile Exam Glove, Medium, Pink, 14-6NPNK4, 250/Box
Catalog No. 503044186
Encompass_Preferred
Exam Glove McKesson Pink Nitrile Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated McKesson Pink Nitrile exam gloves are ambidextrous and nonsterile The gloves are durable and reduce tears and punctures Textured fingertips improve ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard cuff length, and a beaded cuff to prevent rolling They are not approved for chemo Single use powder-free gloves prevent hand residue and reduce irritation A conformed fit provides long-lasting comfort These gloves are pink in support of breast cancer research and awareness
Catalog No. 50-304-4186 Supplier Med Vet International Supplier No. 1065402BX
Exam Glove McKesson Pink Nitrile Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated McKesson Pink Nitrile exam gloves are ambidextrous and nonsterile The gloves are durable and reduce tears and punctures Textured fingertips improve ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard cuff length, and a beaded cuff to prevent rolling They are not approved for chemo Single use powder-free gloves prevent hand residue and reduce irritation A conformed fit provides long-lasting comfort These gloves are pink in support of breast cancer research and awareness