Learn More
Med Vet International Exam Glove MICROFLEX MidKnight™ Touch 93-734 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Black Not Rated, 93732080, 100/Box
Catalog No. 503043289
Encompass_Preferred
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9?-inch length Medium size is for those whose palm circumference is between 7½ and 8 inches
Catalog No. 50-304-3289 Supplier Med Vet International Supplier No. 1182923BX
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9?-inch length Medium size is for those whose palm circumference is between 7½ and 8 inches