Learn More
Med Vet International Exam Glove PremierPro™ Plus Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 5063, 200/Box
Catalog No. 503045099
Encompass_Preferred
Fingertip textured; fingertip thickness is 0.09 mm (3.5 mil) Complies with standard ASTM 6978 (Assessment of Medical Exam Gloves for Resistance to Permeation by Chemotherapy Drugs) Meets USP 800 Personal Protection Equipment requirement for use of Chemotherapy Gloves when handling Hazardous Drugs Nonsterile Beaded cuff Ambidextrous Like all medical exam gloves, these are powder free Medium
Catalog No. 50-304-5099 Supplier Med Vet International Supplier No. 897350BX
Fingertip textured; fingertip thickness is 0.09 mm (3.5 mil) Complies with standard ASTM 6978 (Assessment of Medical Exam Gloves for Resistance to Permeation by Chemotherapy Drugs) Meets USP 800 Personal Protection Equipment requirement for use of Chemotherapy Gloves when handling Hazardous Drugs Nonsterile Beaded cuff Ambidextrous Like all medical exam gloves, these are powder free Medium