Learn More
Med Vet International Exam Glove STERLING SG Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Silver Chemo Tested, 41659, 250/Box
Catalog No. 503042847
Encompass_Preferred
Medium, silver gloves Cleared for use with chemotherapy drugs Halyard Health's manufacturing process Boosts the tensile strength of nitrile to increase its barrier protection qualities The process also allows a reduction in glove thickness thereby reducing the force required to stretch the material creating a nitrile glove with the dexterity and tactile sensitivity of natural rubber latex STERLING SG* Nitrile exam gloves come in space-saving 250-count dispenser boxes SMARTPULL* dispensing incorporates two separate openings on the box The first, smaller opening is used when the box is full and helps control dispensing to lessen waste from dropped gloves or multiple dispensing When the box is half empty, the second, larger opening allows easier access to the gloves
Catalog No. 50-304-2847 Supplier Med Vet International Supplier No. 981297BX
Medium, silver gloves Cleared for use with chemotherapy drugs Halyard Health's manufacturing process Boosts the tensile strength of nitrile to increase its barrier protection qualities The process also allows a reduction in glove thickness thereby reducing the force required to stretch the material creating a nitrile glove with the dexterity and tactile sensitivity of natural rubber latex STERLING SG* Nitrile exam gloves come in space-saving 250-count dispenser boxes SMARTPULL* dispensing incorporates two separate openings on the box The first, smaller opening is used when the box is full and helps control dispensing to lessen waste from dropped gloves or multiple dispensing When the box is half empty, the second, larger opening allows easier access to the gloves