Learn More
Med Vet International Exam Glove Halyard™ Lavender™ Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Lavender Not Rated, 52818, 2500/Case
Catalog No. 503043676
Encompass_Preferred
Lightweight and comfortable Halyard Lavender nitrile gloves are suitable for tasks such as oral care, physical exams and dispensing of medications Flexible material allows for full dexterity and a free range of finger movement A powder- and latex-free design reduces reactions in sensitive skin The ambidextrous design fits either hand Textured fingertips provide a sure grip on containers and implements These Halyard exam gloves come in a Smartpull dispenser box with two openings
Catalog No. 50-304-3676 Supplier Med Vet International Supplier No. 678086CS
Lightweight and comfortable Halyard Lavender nitrile gloves are suitable for tasks such as oral care, physical exams and dispensing of medications Flexible material allows for full dexterity and a free range of finger movement A powder- and latex-free design reduces reactions in sensitive skin The ambidextrous design fits either hand Textured fingertips provide a sure grip on containers and implements These Halyard exam gloves come in a Smartpull dispenser box with two openings