Learn More
Med Vet International Exam Glove McKesson Confiderm LDC Small NonSterile Nitrile Standard Cuff Length Fully Textured Black Chemo Tested / Fentanyl Tested, 14-6N52C, 250/Box
Catalog No. 503044151
Encompass_Preferred
Low dermatitis potential; ideal for individuals with skin sensitivities to regular nitrile gloves Tested to resist permeation of 15 chemotherapy drugs, plus fentanyl Exterior's fully textured finish promotes slip resistance in wet or dry conditions Thin with high-tactile sensitivity Excellent conformability provides superior fit and extended user comfort Durability and tensile strength thanks to nitrile construction Fingertip thickness: 0.07mm (2.8 mil) Chlorinated internal surface Small
Catalog No. 50-304-4151 Supplier Med Vet International Supplier No. 1159333BX
Low dermatitis potential; ideal for individuals with skin sensitivities to regular nitrile gloves Tested to resist permeation of 15 chemotherapy drugs, plus fentanyl Exterior's fully textured finish promotes slip resistance in wet or dry conditions Thin with high-tactile sensitivity Excellent conformability provides superior fit and extended user comfort Durability and tensile strength thanks to nitrile construction Fingertip thickness: 0.07mm (2.8 mil) Chlorinated internal surface Small