Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Small, Blue, 14-6N32EC, 1000/Case
Catalog No. 503042940
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit McKesson Confiderm small nitrile exam gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up and becoming loose Powder-free McKesson nitrile exam gloves are free of natural rubber latex
Catalog No. 50-304-2940 Supplier Med Vet International Supplier No. 1107940CS
Exam Glove McKesson Confiderm 3.0 Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit McKesson Confiderm small nitrile exam gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up and becoming loose Powder-free McKesson nitrile exam gloves are free of natural rubber latex