Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Small, Blue, NonSterile, 14-6N32, 250/Box
Catalog No. 503045355
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips offer outstanding tactile sensitivity These blue Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up Their outstanding conformity offers a proper fit Powder-free, small nitrile exam gloves are wearable on left or right hand They're not made from natural rubber latex to reduce discomfort and irritation
Catalog No. 50-304-5355 Supplier Med Vet International Supplier No. 957801BX
Exam Glove McKesson Confiderm 3.0 Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips offer outstanding tactile sensitivity These blue Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up Their outstanding conformity offers a proper fit Powder-free, small nitrile exam gloves are wearable on left or right hand They're not made from natural rubber latex to reduce discomfort and irritation