Learn More
Med Vet International McKesson Pink Nitrile Nitrile Exam Glove, Small, Pink, 14-6NPNK2, 2500/Case
Catalog No. 503043379
Encompass_Preferred
Exam Glove McKesson Pink Nitrile Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated McKesson pink nitrile exam gloves are made durable to reduce tears and punctures The textured fingers improve your ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard length beaded cuff to prevent them from rolling down They're powder-free to eliminate residue on the hand and reduce irritation The conforming fit provides long-lasting comfort These small exam gloves are pink in support of breast cancer research and awareness
Catalog No. 50-304-3379 Supplier Med Vet International Supplier No. 1065401CS
Exam Glove McKesson Pink Nitrile Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated McKesson pink nitrile exam gloves are made durable to reduce tears and punctures The textured fingers improve your ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard length beaded cuff to prevent them from rolling down They're powder-free to eliminate residue on the hand and reduce irritation The conforming fit provides long-lasting comfort These small exam gloves are pink in support of breast cancer research and awareness