Learn More
Med Vet International Exam Glove Purple Nitrile-Xtra™ Small NonSterile Nitrile Extended Cuff Length Textured Fingertips Purple Chemo Tested, 50601, 50/Box
Catalog No. 503044586
Encompass_Preferred
Halyard Purple Nitrile-Xtra Gloves have textured fingertips with a 0.15mm (5.9 mil) thickness Their purple color makes it easy to locate them These 12-inch gloves provide maximum coverage and protection The small purple Nitrile-Xtra gloves have an enhanced grip for support on wet areas The gloves are disposable and meet the requirements of the government for use in chemotherapy
Catalog No. 50-304-4586 Supplier Med Vet International Supplier No. 365065BX
Halyard Purple Nitrile-Xtra Gloves have textured fingertips with a 0.15mm (5.9 mil) thickness Their purple color makes it easy to locate them These 12-inch gloves provide maximum coverage and protection The small purple Nitrile-Xtra gloves have an enhanced grip for support on wet areas The gloves are disposable and meet the requirements of the government for use in chemotherapy