Learn More
Med Vet International Exam Glove Purple Nitrile Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Purple Chemo Tested, 55081, 100/Box
Catalog No. 503045003
Encompass_Preferred
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical gloves, these are powder free Purple Nitrile small exam gloves are 9½ inches in length Textured fingertips offer better grip in both wet and dry conditions These powder-free exam gloves exceed AQL requirements with a rating of 1.0
Catalog No. 50-304-5003 Supplier Med Vet International Supplier No. 365060BX
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical gloves, these are powder free Purple Nitrile small exam gloves are 9½ inches in length Textured fingertips offer better grip in both wet and dry conditions These powder-free exam gloves exceed AQL requirements with a rating of 1.0