Learn More
Med Vet International Exam Glove Pulse Nitrile Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Aqua Blue Chemo Tested, 177102, 200/Box
Catalog No. 503044615
Encompass_Preferred
Pulse Nitrile gloves have been tested for use with chemotherapy drugs Finely textured finger tips create an unequalled tactile sensitivity and excellent gripping properties Small
Catalog No. 50-304-4615 Supplier Med Vet International Supplier No. 764269BX
Pulse Nitrile gloves have been tested for use with chemotherapy drugs Finely textured finger tips create an unequalled tactile sensitivity and excellent gripping properties Small