Learn More
Med Vet International Exam Glove Micro-Touch Nitrile Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 6034301, 2000/Case
Catalog No. 503043291
Encompass_Preferred
They offer a snug, comfortable fit for better tactile sensitivity The blue, size small nonsterile gloves are sturdy and durable Tested for use with 55 chemicals, and approved for chemotherapy drugs Small size is for those whose palm circumference is between 6½ and 7 inches What is included: 200 gloves per box The Ansell Micro-Touch Nitrile is powder free and has standard cuff length
Catalog No. 50-304-3291 Supplier Med Vet International Supplier No. 703150CS
They offer a snug, comfortable fit for better tactile sensitivity The blue, size small nonsterile gloves are sturdy and durable Tested for use with 55 chemicals, and approved for chemotherapy drugs Small size is for those whose palm circumference is between 6½ and 7 inches What is included: 200 gloves per box The Ansell Micro-Touch Nitrile is powder free and has standard cuff length