Learn More
Med Vet International Exam Glove Aquasoft Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 43933, 3000/Case
Catalog No. 503045256
Encompass_Preferred
Higher count per box saves space and reduces waste Nitrile formulation offers comfort and tactile sensitivity comparable to latex without the risk of latex skin irritation and allergies Like all medical exam gloves, these gloves are powder free Small
Catalog No. 50-304-5256 Supplier Med Vet International Supplier No. 975529CS
Higher count per box saves space and reduces waste Nitrile formulation offers comfort and tactile sensitivity comparable to latex without the risk of latex skin irritation and allergies Like all medical exam gloves, these gloves are powder free Small