Learn More
Med Vet International Exam Glove Micro-Touch NitraFree™ Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Chemo Tested, 6034511, 1000/Case
Catalog No. 503043282
Encompass_Preferred
Ansell's Micro-Touch NitraFree exam gloves protect against latex Type I allergies and reduce chemical Type IV allergies They're ideal for performing exams or caring for ill family members at home These lightweight gloves offer a secure fit and boast textured fingers and a standard cuff length Use these gloves for protection against harsh chemicals and cleaners This disposable product is safe to use with chemotherapy medication These small nitrile exam gloves are pink in support of breast cancer research Small size is for those whose palm circumference is between 6½ and 7 inches
Catalog No. 50-304-3282 Supplier Med Vet International Supplier No. 693950CS
Ansell's Micro-Touch NitraFree exam gloves protect against latex Type I allergies and reduce chemical Type IV allergies They're ideal for performing exams or caring for ill family members at home These lightweight gloves offer a secure fit and boast textured fingers and a standard cuff length Use these gloves for protection against harsh chemicals and cleaners This disposable product is safe to use with chemotherapy medication These small nitrile exam gloves are pink in support of breast cancer research Small size is for those whose palm circumference is between 6½ and 7 inches