Learn More
Med Vet International McKesson Confiderm STR Nitrile Exam Glove, Small, Blue, 14-6NSTR2, 400/Case
Catalog No. 503043825
Encompass_Preferred
Exam Glove McKesson Confiderm STR Small Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These small McKesson exam gloves made of nitrile resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility The powder-free construction of these blue exam gloves eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies
Catalog No. 50-304-3825 Supplier Med Vet International Supplier No. 1065405CS
Exam Glove McKesson Confiderm STR Small Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These small McKesson exam gloves made of nitrile resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility The powder-free construction of these blue exam gloves eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies