Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Extra Large, Blue, 14-6N38EC, 100/Box
Catalog No. 503044698
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile exam gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug, conforming fit McKesson Confiderm gloves are ambidextrous for convenient wear Beaded cuffs prevent the gloves from rolling up and becoming loose The extra-large, powder-free McKesson nitrile exam gloves are made without natural rubber latex
Catalog No. 50-304-4698 Supplier Med Vet International Supplier No. 1107943BX
Exam Glove McKesson Confiderm 3.0 X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile exam gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug, conforming fit McKesson Confiderm gloves are ambidextrous for convenient wear Beaded cuffs prevent the gloves from rolling up and becoming loose The extra-large, powder-free McKesson nitrile exam gloves are made without natural rubber latex