Learn More
Med Vet International McKesson Pink Nitrile Nitrile Exam Glove, Extra Large, Pink, 14-6NPNK8, 230/Box
Catalog No. 503043838
Encompass_Preferred
Exam Glove McKesson Pink Nitrile X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated These gloves are durable to reduce tears and punctures They're rubber-free and non-allergic Textured fingers improve the ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard length and a beaded cuff to prevent them from rolling down They're powder-free to eliminate residue on the hand and reduce irritation The ambidextrous gloves have a conforming fit to provide long-lasting comfort The single-use gloves are nonsterile and not chemo approved These extra-large gloves are pink in support of breast cancer research and awareness
Catalog No. 50-304-3838 Supplier Med Vet International Supplier No. 1065404BX
Exam Glove McKesson Pink Nitrile X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated These gloves are durable to reduce tears and punctures They're rubber-free and non-allergic Textured fingers improve the ability to grasp and hold onto items McKesson Pink Nitrile exam gloves have a standard length and a beaded cuff to prevent them from rolling down They're powder-free to eliminate residue on the hand and reduce irritation The ambidextrous gloves have a conforming fit to provide long-lasting comfort The single-use gloves are nonsterile and not chemo approved These extra-large gloves are pink in support of breast cancer research and awareness