Learn More
Med Vet International McKesson Confiderm 3.5C Nitrile Exam Glove, Extra Large, Blue, NonSterile, 14-6980C, 180/Box
Catalog No. 503043623
Encompass_Preferred
Exam Glove McKesson Confiderm 3.5C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous, offering a quick and easy fit on either hand Confiderm 3.5C gloves are powder- and latex-free, reducing chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile, extra-large exam gloves meet requirements for handling chemicals such as chemotherapy drugs
Catalog No. 50-304-3623 Supplier Med Vet International Supplier No. 767197BX
Exam Glove McKesson Confiderm 3.5C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous, offering a quick and easy fit on either hand Confiderm 3.5C gloves are powder- and latex-free, reducing chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile, extra-large exam gloves meet requirements for handling chemicals such as chemotherapy drugs