Learn More
Med Vet International McKesson Confiderm 3.8 Nitrile Exam Glove, Extra Large, Blue, NonSterile, 14-690, 100/Box
Catalog No. 503044133
Encompass_Preferred
Exam Glove McKesson Confiderm 3.8 X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated McKesson Confiderm gloves have textured fingertips that offer enhanced grip and finger movement The beaded cuffs help keep gloves in place These blue nitrile exam gloves are not made with natural rubber latex to reduce irritation These extra-large powder-free, nonsterile gloves are wearable on the left or right hand These single-use gloves conform well to hand shapes and have a standard cuff length to keep users comfortable They are not approved for use with chemotherapy Extra large
Catalog No. 50-304-4133 Supplier Med Vet International Supplier No. 921612BX
Exam Glove McKesson Confiderm 3.8 X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated McKesson Confiderm gloves have textured fingertips that offer enhanced grip and finger movement The beaded cuffs help keep gloves in place These blue nitrile exam gloves are not made with natural rubber latex to reduce irritation These extra-large powder-free, nonsterile gloves are wearable on the left or right hand These single-use gloves conform well to hand shapes and have a standard cuff length to keep users comfortable They are not approved for use with chemotherapy Extra large