Learn More
Med Vet International McKesson Confiderm 4.5C Nitrile Exam Glove, Extra Large, Blue, NonSterile, 14-660C, 100/Box
Catalog No. 503045097
Encompass_Preferred
Exam Glove McKesson Confiderm 4.5C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C extra-large gloves in place during use Powder-free and latex-free to help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These blue exam gloves are wearable on either hand
Catalog No. 50-304-5097 Supplier Med Vet International Supplier No. 921605BX
Exam Glove McKesson Confiderm 4.5C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C extra-large gloves in place during use Powder-free and latex-free to help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These blue exam gloves are wearable on either hand