Learn More
Med Vet International McKesson Confiderm STR Nitrile Exam Glove, Extra Large, Blue, 14-6NSTR8, Pair
Catalog No. 503042929
Encompass_Preferred
Exam Glove McKesson Confiderm STR X-Large Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These extra-large nitrile exam gloves resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility Powder-free construction eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies
Catalog No. 50-304-2929 Supplier Med Vet International Supplier No. 1065408PR
Exam Glove McKesson Confiderm STR X-Large Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These extra-large nitrile exam gloves resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility Powder-free construction eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies