Learn More
Med Vet International McKesson Confiderm 4.5C Nitrile Exam Glove, Extra Small, Blue, NonSterile, 14-652C, 100/Box
Catalog No. 503043753
Encompass_Preferred
Exam Glove McKesson Confiderm 4.5C X-Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C x-small gloves in place during use They are powder-free and latex-free to help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These blue exam gloves are wearable on either hand
Catalog No. 50-304-3753 Supplier Med Vet International Supplier No. 921601BX
Exam Glove McKesson Confiderm 4.5C X-Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C x-small gloves in place during use They are powder-free and latex-free to help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These blue exam gloves are wearable on either hand