Learn More
Med Vet International Exam Glove Halyard™ Lavender™ X-Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Lavender Not Rated, 52816, 2500/Case
Catalog No. 503044195
Encompass_Preferred
Lightweight and comfortable Halyard Lavender nitrile gloves are suitable for tasks such as oral care, physical exams and dispensing of medications Flexible material allows for full dexterity and a free range of finger movement A powder- and latex-free design reduces reactions in sensitive skin The ambidextrous design fits either hand Textured fingertips with a 0.07 mm thickness provide a sure grip on containers and implements These nonsterile extra-small nitrile powder-free gloves come in a Smartpull dispenser box with two openings
Catalog No. 50-304-4195 Supplier Med Vet International Supplier No. 678083CS
Lightweight and comfortable Halyard Lavender nitrile gloves are suitable for tasks such as oral care, physical exams and dispensing of medications Flexible material allows for full dexterity and a free range of finger movement A powder- and latex-free design reduces reactions in sensitive skin The ambidextrous design fits either hand Textured fingertips with a 0.07 mm thickness provide a sure grip on containers and implements These nonsterile extra-small nitrile powder-free gloves come in a Smartpull dispenser box with two openings