Learn More
Med Vet International McKesson Vinyl Exam Glove, Small, Clear, NonSterile, 14-114, 1000/Case
Catalog No. 503043016
Encompass_Preferred
Exam Glove McKesson Small NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson exam gloves are made of tear and puncture-resistant vinyl The smooth finish throughout lets you apply and remove them easily McKesson vinyl exam gloves stretch to provide a conformed fit that improves tactile sensitivity, dexterity and comfort Like all medical gloves, these are powder free Beaded cuff eliminates roll-down and drips They're latex-free, allowing for use by those with latex allergies The ambidextrous construction permits use on the left and right hand Small
Catalog No. 50-304-3016 Supplier Med Vet International Supplier No. 354438CS
Exam Glove McKesson Small NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson exam gloves are made of tear and puncture-resistant vinyl The smooth finish throughout lets you apply and remove them easily McKesson vinyl exam gloves stretch to provide a conformed fit that improves tactile sensitivity, dexterity and comfort Like all medical gloves, these are powder free Beaded cuff eliminates roll-down and drips They're latex-free, allowing for use by those with latex allergies The ambidextrous construction permits use on the left and right hand Small