Learn More
Med Vet International McKesson Vinyl Exam Glove, Large, Clear, NonSterile, 14-118, 1000/Case
Catalog No. 503044708
Encompass_Preferred
Exam Glove McKesson Large NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson exam gloves are made of tear and puncture-resistant vinyl The smooth finish throughout lets you apply and remove them easily McKesson vinyl exam gloves stretch to provide a conformed fit that improves tactile sensitivity, dexterity and comfort The powder-free construction leaves hands residue free Beaded cuff eliminates roll-down and drips The ambidextrous construction allows for use on the left and right hand Large
Catalog No. 50-304-4708 Supplier Med Vet International Supplier No. 354440CS
Exam Glove McKesson Large NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson exam gloves are made of tear and puncture-resistant vinyl The smooth finish throughout lets you apply and remove them easily McKesson vinyl exam gloves stretch to provide a conformed fit that improves tactile sensitivity, dexterity and comfort The powder-free construction leaves hands residue free Beaded cuff eliminates roll-down and drips The ambidextrous construction allows for use on the left and right hand Large