Learn More
Med Vet International Exam Glove Micro-Touch Elite Large NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated, 3093, 1000/Case
Catalog No. 503044019
Encompass_Preferred
Ivory exam gloves have a formulation created for a more comfortable and secure fit than traditional vinyl gloves Smooth finish offers enhanced tactile sensitivity Like all medical gloves, these Micro-Touch Elite gloves are powder free Fingertip thickness is 5.9 mil Large size is for those whose palm circumference is between 8½ and 9 inches
Catalog No. 50-304-4019 Supplier Med Vet International Supplier No. 453105CS
Ivory exam gloves have a formulation created for a more comfortable and secure fit than traditional vinyl gloves Smooth finish offers enhanced tactile sensitivity Like all medical gloves, these Micro-Touch Elite gloves are powder free Fingertip thickness is 5.9 mil Large size is for those whose palm circumference is between 8½ and 9 inches