Learn More
Med Vet International McKesson Stretch Vinyl Exam Glove, Large, Ivory, NonSterile, 14-818, 1000/Case
Catalog No. 503045435
Encompass_Preferred
Exam Glove McKesson Large NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The ivory, vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the stretch vinyl exam gloves with ease The large gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down
Catalog No. 50-304-5435 Supplier Med Vet International Supplier No. 409744CS
Exam Glove McKesson Large NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The ivory, vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the stretch vinyl exam gloves with ease The large gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down