Learn More
Med Vet International Exam Glove Micro-Touch Elite Medium NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated, 3092, 100/Box
Catalog No. 503044612
Encompass_Preferred
Ivory exam gloves have a formulation created for a more comfortable and secure fit than traditional vinyl gloves Smooth finish offers enhanced tactile sensitivity Like all medical gloves, these Micro-Touch Elite gloves are powder free Fingertip thickness is 5.9 mil Medium size fits those whose palm circumference is between 7½ and 8 inches
Catalog No. 50-304-4612 Supplier Med Vet International Supplier No. 453104BX
Ivory exam gloves have a formulation created for a more comfortable and secure fit than traditional vinyl gloves Smooth finish offers enhanced tactile sensitivity Like all medical gloves, these Micro-Touch Elite gloves are powder free Fingertip thickness is 5.9 mil Medium size fits those whose palm circumference is between 7½ and 8 inches