Learn More
Med Vet International Exam Glove Halyard™ Medium NonSterile Vinyl Standard Cuff Length Smooth White Not Rated, 50032, 1000/Case
Catalog No. 503045522
Encompass_Preferred
Polymer coating improves donning but fingertips are smooth Ambidextrous for use across a variety of glove applications 9-1/2 inches long Medium
Catalog No. 50-304-5522 Supplier Med Vet International Supplier No. 447638CS
 Restricted Item
Add to Cart
Add to Cart
Polymer coating improves donning but fingertips are smooth Ambidextrous for use across a variety of glove applications 9-1/2 inches long Medium