Learn More
Med Vet International McKesson Stretch Vinyl Exam Glove, Medium, Ivory, NonSterile, 14-816, 1000/Case
Catalog No. 503044803
Encompass_Preferred
Exam Glove McKesson Medium NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the McKesson medium exam gloves with ease The ivory gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down
Catalog No. 50-304-4803 Supplier Med Vet International Supplier No. 409743CS
Exam Glove McKesson Medium NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the McKesson medium exam gloves with ease The ivory gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down