Learn More
Med Vet International McKesson Non-Sterile, Powder-Free Vinyl Exam Gloves, Medium, Standard Cuff Length, Smooth Clear, 14-136, 1500/Case
Catalog No. 503043456
Encompass_Preferred
Exam Glove McKesson Medium NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hands The smooth inner and outer surfaces create a comfortable fit for a natural touch These medium size powder-free, clear exam gloves are not made for use with chemotherapy
Catalog No. 50-304-3456 Supplier Med Vet International Supplier No. 832682CS
Exam Glove McKesson Medium NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hands The smooth inner and outer surfaces create a comfortable fit for a natural touch These medium size powder-free, clear exam gloves are not made for use with chemotherapy