Learn More
Med Vet International McKesson Vinyl Exam Glove, Small, Clear, NonSterile, 14-134, 150/Box
Catalog No. 503043321
Encompass_Preferred
Exam Glove McKesson Small NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hands The smooth inner and outer surfaces create a comfortable fit for a natural touch These size small powder-free exam gloves are not made for use with chemotherapy
Catalog No. 50-304-3321 Supplier Med Vet International Supplier No. 832681BX
Exam Glove McKesson Small NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hands The smooth inner and outer surfaces create a comfortable fit for a natural touch These size small powder-free exam gloves are not made for use with chemotherapy