Learn More
Med Vet International McKesson Vinyl Exam Glove, Extra Large, Clear, NonSterile, 14-140, 1300/Case
Catalog No. 503043944
Encompass_Preferred
Exam Glove McKesson X-Large NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hand The smooth inner and outer surfaces create a comfortable fit for a natural touch These extra-large size powder-free exam gloves are not approved for use with chemotherapy
Catalog No. 50-304-3944 Supplier Med Vet International Supplier No. 833078CS
Exam Glove McKesson X-Large NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated McKesson powder-free vinyl exam gloves protect your hands during patient exams This disposable medical product is not sterile Use a pair of these clear exam gloves for one patient exam only The gloves have standard cuffs and rolled edges for easy on and off Wear McKesson vinyl exam gloves on either the right or left hand The smooth inner and outer surfaces create a comfortable fit for a natural touch These extra-large size powder-free exam gloves are not approved for use with chemotherapy