Learn More
Med Vet International McKesson Stretch Vinyl Exam Glove, Extra Large, Ivory, NonSterile, 14-820, 1000/Case
Catalog No. 503043570
Encompass_Preferred
Exam Glove McKesson X-Large NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson stretch vinyl exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the McKesson vinyl exam gloves with ease The ivory, x-large exam gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down
Catalog No. 50-304-3570 Supplier Med Vet International Supplier No. 409745CS
Exam Glove McKesson X-Large NonSterile Stretch Vinyl Standard Cuff Length Smooth Ivory Not Rated McKesson stretch vinyl exam gloves stretch to provide a personalized, comfortable fit and improved tactile sensitivity The vinyl material resists tears and punctures for reliable performance They are latex-free for use on skin with latex allergies The smooth finish lets you apply and remove the McKesson vinyl exam gloves with ease The ivory, x-large exam gloves feature a residue-free interior for added comfort They have a standard beaded cuff to reduce roll-down