Learn More
Med Vet International McKesson Confiderm 4.5C Nitrile Exam Glove, Large, Blue, NonSterile, 14-658C, 1000/Case
Catalog No. 503043755
Encompass_Preferred
Exam Glove McKesson Confiderm 4.5C Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C gloves in place during use The powder-free and latex-free gloves help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These large size exam gloves are wearable on either hand These gloves are chemo tested
Catalog No. 50-304-3755 Supplier Med Vet International Supplier No. 921604CS
Exam Glove McKesson Confiderm 4.5C Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C gloves in place during use The powder-free and latex-free gloves help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These large size exam gloves are wearable on either hand These gloves are chemo tested