Learn More
Med Vet International McKesson Confiderm 6.5CX Extended Cuff Nitrile Extended Cuff Length Exam Glove, Large, Blue, NonSterile, 14-678C, 100/Box
Catalog No. 503045087
Encompass_Preferred
Exam Glove McKesson Confiderm 6.5CX Large NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The size blue, nitrile McKesson exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended, beaded cuff for easy applying and added forearm protection Like all medical gloves, these are powder free Latex-free, extended cuff exam gloves are safe for use with all skin types Large
Catalog No. 50-304-5087 Supplier Med Vet International Supplier No. 921608BX
Exam Glove McKesson Confiderm 6.5CX Large NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The size blue, nitrile McKesson exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended, beaded cuff for easy applying and added forearm protection Like all medical gloves, these are powder free Latex-free, extended cuff exam gloves are safe for use with all skin types Large