Learn More
Med Vet International Exam Glove StarMed Rose Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Not Rated, SMNR203, 200/Box
Catalog No. 503045137
Encompass_Preferred
StarMed Rose Nitrile with Aloe & Vitamin E has what it takes to brighten any environment, from ER to exam room An aloe and vitamin E coating soothes hands and helps prevent skin irritation, while StarMed Rose’s formulation provides enhanced protection, comfort and tactile sensitivity This glove also features Smart Dispense Technology, Single Glove Delivery packaging, which reduces wrinkles and allows for greater efficiency
Catalog No. 50-304-5137 Supplier Med Vet International Supplier No. 976525BX
StarMed Rose Nitrile with Aloe & Vitamin E has what it takes to brighten any environment, from ER to exam room An aloe and vitamin E coating soothes hands and helps prevent skin irritation, while StarMed Rose’s formulation provides enhanced protection, comfort and tactile sensitivity This glove also features Smart Dispense Technology, Single Glove Delivery packaging, which reduces wrinkles and allows for greater efficiency