Learn More
Med Vet International McKesson Confiderm 3.5C Nitrile Exam Glove, Medium, Blue, 14-6976C, 200/Box
Catalog No. 503044364
Encompass_Preferred
Exam Glove McKesson Confiderm 3.5C Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous, offering a quick and easy fit on either hand Confiderm 3.5C medium, blue gloves are powder- and latex-free, reducing chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile exam gloves meet requirements for handling chemicals such as chemotherapy drugs Medium
Catalog No. 50-304-4364 Supplier Med Vet International Supplier No. 765875BX
Exam Glove McKesson Confiderm 3.5C Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous, offering a quick and easy fit on either hand Confiderm 3.5C medium, blue gloves are powder- and latex-free, reducing chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile exam gloves meet requirements for handling chemicals such as chemotherapy drugs Medium