Learn More
Med Vet International McKesson Confiderm 6.5CX Extended Cuff Nitrile Extended Cuff Length Exam Glove, Medium, Blue, NonSterile, 14-676C, 1000/Case
Catalog No. 503045209
Encompass_Preferred
Exam Glove McKesson Confiderm 6.5CX Medium NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick McKesson nitrile exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended cuff for easy applying and added forearm protection Like all medical gloves, these are powder free Latex-free, extended cuff gloves are safe for use with all skin types Medium
Catalog No. 50-304-5209 Supplier Med Vet International Supplier No. 921607CS
Exam Glove McKesson Confiderm 6.5CX Medium NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick McKesson nitrile exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended cuff for easy applying and added forearm protection Like all medical gloves, these are powder free Latex-free, extended cuff gloves are safe for use with all skin types Medium