Learn More
Med Vet International McKesson Confiderm 6.5CX Extended Cuff Nitrile Extended Cuff Length Exam Glove, Small, Blue, NonSterile, 14-674C, 1000/Case
Catalog No. 503044077
Encompass_Preferred
Exam Glove McKesson Confiderm 6.5CX Small NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick blue exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items The McKesson Confiderm gloves feature a long extended, beaded cuff for easy applying and added forearm protection They're powder-free to eliminate hand residue The size small, latex-free gloves are safe for use with all skin types
Catalog No. 50-304-4077 Supplier Med Vet International Supplier No. 921606CS
Exam Glove McKesson Confiderm 6.5CX Small NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick blue exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items The McKesson Confiderm gloves feature a long extended, beaded cuff for easy applying and added forearm protection They're powder-free to eliminate hand residue The size small, latex-free gloves are safe for use with all skin types