Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Extra Large, Blue, Non-Sterile, 14-6N38, 2300/Case
Catalog No. 503045389
Encompass_Preferred
Textured fingertips offer outstanding tactile sensitivity. Fingertip thickness is 0.08 mm (3.1 mil). These blue Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up. Their outstanding conformity offers a proper fit. Like all medical gloves, these exam gloves are powder free. Measure your palm width along the palmar crease: Extra large size will fit those whose palm width is 4 – 4.4 inches.
Catalog No. 50-304-5389 Supplier Med Vet International Supplier No. 957804CS
Textured fingertips offer outstanding tactile sensitivity. Fingertip thickness is 0.08 mm (3.1 mil). These blue Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up. Their outstanding conformity offers a proper fit. Like all medical gloves, these exam gloves are powder free. Measure your palm width along the palmar crease: Extra large size will fit those whose palm width is 4 – 4.4 inches.