Learn More
Med Vet International McKesson Confiderm 6.5CX Extended Cuff Nitrile Extended Cuff Length Exam Glove, Extra Large, Blue, NonSterile, 14-680C, 1000/Case
Catalog No. 503043798
Encompass_Preferred
Exam Glove McKesson Confiderm 6.5CX X-Large NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick McKesson exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended, beaded cuff for easy applying and more coverage Like all medical gloves, these are powder free Latex-free gloves are safe for use with all skin types Extra large
Catalog No. 50-304-3798 Supplier Med Vet International Supplier No. 921609CS
Exam Glove McKesson Confiderm 6.5CX X-Large NonSterile Nitrile Extended Cuff Length Textured Fingertips Blue Chemo Tested The extra-thick McKesson exam gloves are resistant to tears and punctures Textured fingertips allow you to collect, retain and use items Long extended, beaded cuff for easy applying and more coverage Like all medical gloves, these are powder free Latex-free gloves are safe for use with all skin types Extra large