Learn More
Med Vet International McKesson Confiderm 6.8C Nitrile Exam Glove, Extra Large, Blue, NonSterile, 14-6N68C, 1000/Case
Catalog No. 503043272
Encompass_Preferred
Exam Glove McKesson Confiderm 6.8C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested / Fentanyl Tested Textured gloves protect from toxic chemicals often encountered when dealing with fentanyl overdoses Standard length cuff is beaded Tested for use with 32 chemotherapy drugs; meets ASTM standards for resistance to permeation by chemotherapy drugs Tested to resist permeation of fentanyl citrate for up to 240 minutes The interior of the gloves is not coated Extra large
Catalog No. 50-304-3272 Supplier Med Vet International Supplier No. 1163942CS
Exam Glove McKesson Confiderm 6.8C X-Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested / Fentanyl Tested Textured gloves protect from toxic chemicals often encountered when dealing with fentanyl overdoses Standard length cuff is beaded Tested for use with 32 chemotherapy drugs; meets ASTM standards for resistance to permeation by chemotherapy drugs Tested to resist permeation of fentanyl citrate for up to 240 minutes The interior of the gloves is not coated Extra large