Learn More
Med Vet International McKesson Confiderm 4.5C Nitrile Exam Glove, Extra Small, Blue, NonSterile, 14-652C, 1000/Case
Supplier: Med Vet International 921601CS
Exam Glove McKesson Confiderm 4.5C X-Small NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested Beaded cuffs help keep Confiderm 4.5 C x-small gloves in place during use They are powder-free and latex-free to help ensure user comfort McKesson Confiderm gloves are safe for use with chemotherapy drugs Textured fingertips allow for easy gripping, handling and movement of fingers These blue exam gloves are wearable on either hand