Learn More
Refrigiwear Inc. 0679R REGULAR BLACK XLARGE Extreme Freezer Glove
SDP
Catalog No. NC1020408
Encompass_Preferred
Built for hard work in the coldest temperatures, you can put the Extreme Freezer Gloves through the ringer time & time again. With impact protection & just about the most durable construction you'll find, these insulated work gloves protect your hands from hazards on the job as well as harsh -30�F conditions. Keep out extreme winter weather with these pre-curved & ergonomic-fitted work gloves. The knitted fabric back with a synthetic leather palm & silicone grip pattern on the palm keeps your hands warm & your grip secure. Even the strongest winds won't cut through the 200g insulation in these tough impact protection gloves. The entire Extreme Collection, now offering complete head to toe protection, was designed for comfort, style, & the ultimate protection. When the cold warns you to stay at home, take it to the Extreme. Comfort Rating: -30�F 200g AirBlaze® insulation laminated to a tricot lining Knitted fabric back with synthetic leather palm Pre-curved, ergonomic fit Silicon
This item is not returnable. View return policy
Catalog No. NC1020408 Supplier Refrigiwear Inc. Supplier No. 0679RBLKXLG
Built for hard work in the coldest temperatures, you can put the Extreme Freezer Gloves through the ringer time & time again. With impact protection & just about the most durable construction you'll find, these insulated work gloves protect your hands from hazards on the job as well as harsh -30�F conditions. Keep out extreme winter weather with these pre-curved & ergonomic-fitted work gloves. The knitted fabric back with a synthetic leather palm & silicone grip pattern on the palm keeps your hands warm & your grip secure. Even the strongest winds won't cut through the 200g insulation in these tough impact protection gloves. The entire Extreme Collection, now offering complete head to toe protection, was designed for comfort, style, & the ultimate protection. When the cold warns you to stay at home, take it to the Extreme. Comfort Rating: -30�F 200g AirBlaze® insulation laminated to a tricot lining Knitted fabric back with synthetic leather palm Pre-curved, ergonomic fit Silicon