Learn More
MSC Glove Guard LP Clip, 15 lb Safety Breakaway, 1 ea
Catalog No. 5000686146
Encompass
The Glove Guard® clip 1939 is the original glove clip, designed with a flexible, redundant safety breakaway. It significantly reduces glove replacement costs and hand injuries by ensuring workers have their gloves readily available when needed.
  • Made from an engineered grade material that holds its memory
  • Resists flex fatigue
  • Features a 15 lb safety breakaway
  • Interlocking teeth on both ends
  • Non-conductive and non-corrosive
Catalog No. 50-006-86146 Supplier MSC Supplier No. 66346289
The Glove Guard® clip 1939 is the original glove clip, designed with a flexible, redundant safety breakaway. It significantly reduces glove replacement costs and hand injuries by ensuring workers have their gloves readily available when needed.
  • Made from an engineered grade material that holds its memory
  • Resists flex fatigue
  • Features a 15 lb safety breakaway
  • Interlocking teeth on both ends
  • Non-conductive and non-corrosive