Learn More
MSC Glove Guard LP Glove Clip, 15 lb Breakaway, 1 ea
Catalog No. 5000766937
Encompass
The 1939 Glove Guard® clip is the original glove clip, designed to reduce glove replacement costs and hand injuries by ensuring workers have their gloves readily available. It features a flexible, redundant safety breakaway mechanism with a 15 lb safety breakaway.
  • Non-corrosive and non-conductive
  • Made from engineered grade material that maintains its memory
  • Resists flex fatigue
  • Features a 15 lb safety breakaway
  • Interlocking teeth on both ends
  • Available in 12 different colors
Catalog No. 50-007-66937 Supplier MSC Supplier No. 73238990
The 1939 Glove Guard® clip is the original glove clip, designed to reduce glove replacement costs and hand injuries by ensuring workers have their gloves readily available. It features a flexible, redundant safety breakaway mechanism with a 15 lb safety breakaway.
  • Non-corrosive and non-conductive
  • Made from engineered grade material that maintains its memory
  • Resists flex fatigue
  • Features a 15 lb safety breakaway
  • Interlocking teeth on both ends
  • Available in 12 different colors