Learn More
Med Vet International Dynarex Aloetex Latex Gloves w/Aloe, Large, Green, Powder-Free, 1000/Case
Supplier:  Med Vet International 6719
Encompass_Preferred
Dynarex Aloetex Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and comfort. To help moisturize and nourish skin, the Gloves are infused with soothing aloe. The ambidextrous Gloves are designed with textured palms for increased grip and a minimum thickness of 6 mil at the palm and 7 mil at the fingertips for tactile sensitivity. The Gloves feature a polymer lining for improved tear-resistance and beaded cuffs to help ensure easy donning. These Exam Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide range of healthcare facilities and professional environments. Dynarex Aloetex Latex Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These Gloves are coated with skin-soothing aloe to provide optimal comfort.
Catalog No. 50-282-9954